Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aass
Entrez Gene ID
30956
Gene Name
aminoadipate-semialdehyde synthase
Gene Aliases
LKR, LKR/SDH, LOR, LOR/SDH, Lorsdh, SDH
Chromosome Location
Chr.6: 23072173 - 23132986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013930.4 NP_038958.2 12 - 13 1454 118
GenBank mRNA AJ224761.1 - 11 - 12 1345 118
AK098141.1 - 12 - 13 1395 118
AK136216.1 - 12 - 13 1403 118
AK149400.1 - 12 - 13 1379 118
BC005420.1 - 12 - 13 1440 118

View Full Product Details