The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130163.1 | NP_001123635.1 | 13 - 14 | 2267 | 107 |
NM_001130164.1 | NP_001123636.1 | 5 - 6 | 620 | 107 | |
NM_001130165.1 | NP_001123637.1 | 13 - 14 | 2362 | 107 | |
NM_001130166.1 | NP_001123638.1 | 16 - 17 | 2707 | 107 | |
NM_130885.2 | NP_570955.1 | 14 - 15 | 2348 | 107 | |
GenBank mRNA | AF324899.1 | - | 14 - 15 | 2347 | 107 |
AF333985.1 | - | 13 - 14 | 2267 | 107 | |
AK158703.1 | - | 16 - 17 | 2689 | 107 | |
BC003927.1 | - | 7 - 8 | 857 | 107 | |
BC089183.1 | - | 13 - 14 | 2357 | 107 | |
BC098491.1 | - | 15 - 16 | 2609 | 107 |