The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145374.2 | NP_663349.2 | 5 - 6 | 1934 | 80 |
GenBank mRNA | BC020002.1 | - | 5 - 6 | 1932 | 80 |
BC025652.1 | - | 3 - 4 | 1002 | 80 | |
BQ876300.1 | - | 3 - 4 | 489 | 80 |