The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053098.2 | NP_444328.1 | 1 - 2 | 391 | 105 |
GenBank mRNA | AK147141.1 | - | 1 - 2 | 392 | 105 |