Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grhl2
Entrez Gene ID
252973
Gene Name
grainyhead-like 2 (Drosophila)
Gene Aliases
0610015A08Rik, BOM, Tcfcp2l3
Chromosome Location
Chr.15: 37233036 - 37363569 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026496.4 NP_080772.2 1 - 2 365 71
GenBank mRNA AF411213.1 - 1 - 2 98 71
AK002633.1 - 1 - 2 366 71
BC055035.1 - 1 - 2 154 71
BY018142.1 - 1 - 2 311 71
BY703140.1 - 1 - 2 366 71

View Full Product Details