The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030014.2 | NP_001025185.1 | 3 - 4 | 748 | 83 |
GenBank mRNA | AK135457.1 | - | 3 - 4 | 774 | 83 |
BC025519.1 | - | 1 - 2 | 244 | 83 | |
BC099494.1 | - | 3 - 4 | 748 | 83 |