Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pip4k2c
Entrez Gene ID
117150
Gene Name
phosphatidylinositol-5-phosphate 4-kinase, type II, gamma
Gene Aliases
Pip5k2c
Chromosome Location
Chr.10: 127197067 - 127211644 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054097.3 NP_473438.1 4 - 5 615 79
GenBank mRNA AB054591.1 - 4 - 5 558 79
AK148299.1 - 4 - 5 575 79
AK207886.1 - 2 - 3 223 79
BC021383.1 - 4 - 5 548 79

View Full Product Details