The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003913.2 | NP_001003913.1 | 2 - 3 | 354 | 74 |
NM_001171582.1 | NP_001165053.1 | 2 - 3 | 354 | 74 | |
GenBank mRNA | AK181384.1 | - | 1 - 2 | 65 | 73 |
AK211562.1 | - | 1 - 2 | 65 | 74 | |
AK213658.1 | - | 1 - 2 | 65 | 74 | |
BC052042.1 | - | 2 - 3 | 206 | 74 | |
BC058968.1 | - | 2 - 3 | 246 | 74 | |
BC079643.1 | - | 2 - 3 | 207 | 74 |