Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mars
Entrez Gene ID
216443
Gene Name
methionine-tRNA synthetase
Gene Aliases
Metrs, Mtrns
Chromosome Location
Chr.10: 127296221 - 127311786 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003913.2 NP_001003913.1 6 - 7 821 135
NM_001171582.1 NP_001165053.1 6 - 7 821 135
GenBank mRNA BC052042.1 - 6 - 7 673 135
BC058968.1 - 6 - 7 713 135
BC079643.1 - 6 - 7 674 135

View Full Product Details