The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011166.2 | NP_035296.1 | 4 - 5 | 542 | 133 |
GenBank mRNA | AF011384.1 | - | 4 - 5 | 541 | 133 |
AF015563.1 | - | 4 - 5 | 542 | 133 |