The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008865.3 | NP_032891.1 | 4 - 5 | 536 | 85 |
GenBank mRNA | AK005542.1 | - | 4 - 5 | 537 | 85 |
AK005543.1 | - | 4 - 5 | 536 | 85 | |
AK131667.1 | - | 4 - 5 | 537 | 85 | |
AK167483.1 | - | 4 - 5 | 537 | 85 | |
BC099386.1 | - | 4 - 5 | 521 | 85 | |
M14647.1 | - | 4 - 5 | 536 | 85 |