Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kel
Entrez Gene ID
23925
Gene Name
Kell blood group
Gene Aliases
CD238, Ece3
Chromosome Location
Chr.6: 41686329 - 41704334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032540.3 NP_115929.3 2 - 3 283 86
GenBank mRNA AF252870.1 - 2 - 3 255 86
AK028430.1 - 2 - 3 257 86
BC099961.1 - 2 - 3 281 86
BY234859.1 - 2 - 3 284 86

View Full Product Details