Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mid1
Entrez Gene ID
17318
Gene Name
midline 1
Gene Aliases
61B3-R, DXHXS1141, Fxy, Trim18
Chromosome Location
Chr.X: 169685199 - 169990798 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290504.1 NP_001277433.1 - 64 71
GenBank mRNA AF026565.1 - - 64 71

View Full Product Details