Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phex
Entrez Gene ID
18675
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
Gy, HPDR, HPDR1, Hyp, PEX
Chromosome Location
Chr.X: 157162075 - 157416551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011077.2 NP_035207.1 9 - 10 1613 115
GenBank mRNA AK158481.1 - 9 - 10 1640 115
BC137925.1 - 9 - 10 1360 115
BC137926.1 - 9 - 10 1360 115
EF194891.1 - 9 - 10 1147 115
U49908.1 - 9 - 10 1605 115
U73910.1 - 9 - 10 1613 115
U75646.1 - 9 - 10 1194 115

View Full Product Details