The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009311.2 | NP_033337.1 | 5 - 6 | 394 | 66 |
GenBank mRNA | BC117081.1 | - | 5 - 6 | 369 | 66 |
BC119122.1 | - | 5 - 6 | 369 | 66 | |
D17584.1 | - | 5 - 6 | 394 | 66 | |
M68908.1 | - | 4 - 5 | 190 | 66 |