Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vps13b
- Entrez Gene ID
-
666173
- Gene Name
- vacuolar protein sorting 13B (yeast)
- Gene Aliases
-
1810042B05Rik, 2310042E16Rik, 4732488H20, C330002D13Rik, C87206, Coh1, D230005K13, mKIAA0532, mKIAA5032
- Chromosome Location
-
Chr.15: 35369882 - 35931229 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177151.3 |
NP_796125.2 |
52 - 53 |
9869 |
91 |
GenBank mRNA |
AK122302.1 |
- |
13 - 14 |
2440 |
91 |
View Full Product Details