Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map7d2
Entrez Gene ID
78283
Gene Name
MAP7 domain containing 2
Gene Aliases
1600028E09Rik, 2900002G04Rik, 5330432J06Rik, C030036K04Rik, Mtap7d2
Chromosome Location
Chr.X: 159414527 - 159498961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081124.1 NP_001074593.1 7 - 8 998 119
GenBank mRNA AK005554.1 - 3 - 4 515 119
AK013466.1 - 7 - 8 787 119
AK019929.1 - 3 - 4 596 119
AK030316.1 - 3 - 4 500 119
AK043342.1 - 3 - 4 573 119
AK162227.1 - 4 - 5 433 119

View Full Product Details