Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Birc2
Entrez Gene ID
11797
Gene Name
baculoviral IAP repeat-containing 2
Gene Aliases
AW146227, Api1, Api2, Birc3, HIAP1, HIAP2, IAP1, IAP2, MIAP1, MIAP2, MIHB, MIHC, RNF48, cIAP1, cIAP2, mcIAP1
Chromosome Location
Chr.9: 7818226 - 7837133 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007465.2 NP_031491.2 5 - 6 2379 85
GenBank mRNA BC033480.1 - 5 - 6 2393 85
BC145985.1 - 5 - 6 2232 85
L49433.1 - 5 - 6 2074 85
U88909.1 - 5 - 6 2380 85

View Full Product Details