Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp7
Entrez Gene ID
17393
Gene Name
matrix metallopeptidase 7
Gene Aliases
MAT
Chromosome Location
Chr.9: 7692111 - 7699416 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010810.4 NP_034940.2 4 - 5 640 67
GenBank mRNA AK144515.1 - 4 - 5 658 67
BC119057.1 - 4 - 5 640 67
BC120655.1 - 4 - 5 640 67

View Full Product Details