Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcsk7
Entrez Gene ID
18554
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
AA959856, PC7, SPC7
Chromosome Location
Chr.9: 45906569 - 45929726 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281934.1 NP_001268863.1 12 - 13 2409 63
NM_008794.2 NP_032820.2 12 - 13 2145 63
GenBank mRNA AK148416.1 - 12 - 13 1730 63
AK151980.1 - 12 - 13 1732 63
AK165389.1 - 12 - 13 1751 63
BC006730.1 - 11 - 12 1646 63
BC007322.1 - 12 - 13 2008 63
U48830.1 - 12 - 13 1717 63

View Full Product Details