Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirt1
Entrez Gene ID
93759
Gene Name
sirtuin 1
Gene Aliases
AA673258, SIR2L1, Sir2, Sir2a, Sir2alpha
Chromosome Location
Chr.10: 63319005 - 63339061 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159589.1 NP_001153061.1 3 - 4 865 96
NM_019812.2 NP_062786.1 4 - 5 979 96
GenBank mRNA AF214646.1 - 4 - 5 964 96
AK144203.1 - 3 - 4 868 96
AY377984.1 - 4 - 5 964 96
BC152314.1 - 4 - 5 459 96

View Full Product Details