The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172162.3 | NP_751894.2 | 6 - 7 | 803 | 88 |
GenBank mRNA | AK049348.1 | - | 6 - 7 | 765 | 88 |
AK049633.1 | - | 6 - 7 | 964 | 88 | |
BC029091.1 | - | 5 - 6 | 786 | 88 | |
CT010239.1 | - | 5 - 6 | 738 | 88 |