Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc37a4
Entrez Gene ID
14385
Gene Name
solute carrier family 37 (glucose-6-phosphate transporter), member 4
Gene Aliases
G6PT, G6pt1, GSD-1b, mG6PT
Chromosome Location
Chr.9: 44396857 - 44402968 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293630.1 NP_001280559.1 3 - 4 941 107
NM_001293631.1 NP_001280560.1 3 - 4 654 107
NM_008063.2 NP_032089.2 2 - 3 681 107
GenBank mRNA AK003620.1 - 2 - 3 504 107
AK147849.1 - 3 - 4 611 107
AK155339.1 - 3 - 4 938 107
BC075655.1 - 2 - 3 681 107

View Full Product Details