Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Treh
Entrez Gene ID
58866
Gene Name
trehalase (brush-border membrane glycoprotein)
Gene Aliases
2210412M19Rik
Chromosome Location
Chr.9: 44673233 - 44686305 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277847.1 NP_001264776.1 6 - 7 650 94
NM_001277851.1 NP_001264780.1 6 - 7 654 94
NM_021481.3 NP_067456.1 7 - 8 755 94
GenBank mRNA AF136944.1 - 7 - 8 750 94
AK002566.1 - 6 - 7 653 94
AK008912.1 - 7 - 8 740 94
BC019214.1 - 6 - 7 645 94

View Full Product Details