Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlrp5
Entrez Gene ID
23968
Gene Name
NLR family, pyrin domain containing 5
Gene Aliases
Mater, Nalp5, Op1, PAN11
Chromosome Location
Chr.7: 23385897 - 23441921 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF074018.1 - 9 - 10 2350 105
AK087774.1 - 9 - 10 2372 105
AY196361.1 - 9 - 10 2350 105
AY196362.1 - 9 - 10 2350 105
AY329484.1 - 8 - 9 2302 105
AY329485.1 - 9 - 10 2350 105
AY329486.1 - 9 - 10 2350 105
AY329487.1 - 8 - 9 2302 105
AY329489.1 - 9 - 10 2350 105
AY329490.1 - 9 - 10 2350 105
BC053384.1 - 10 - 11 2417 105

View Full Product Details