The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008607.2 | NP_032633.1 | 9 - 10 | 1359 | 76 |
GenBank mRNA | AK150728.1 | - | 9 - 10 | 1346 | 76 |
AK151626.1 | - | 9 - 10 | 1346 | 76 | |
AK152507.1 | - | 9 - 10 | 1345 | 76 | |
AK152916.1 | - | 9 - 10 | 1346 | 76 | |
AK153458.1 | - | 9 - 10 | 1344 | 76 | |
AK155188.1 | - | 9 - 10 | 1345 | 76 | |
AK159921.1 | - | 9 - 10 | 1346 | 76 | |
AK160819.1 | - | 9 - 10 | 1344 | 76 | |
BC125320.1 | - | 9 - 10 | 1359 | 76 | |
BC125322.1 | - | 9 - 10 | 1359 | 76 | |
X66473.1 | - | 9 - 10 | 1328 | 76 |