The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023431.5 | NP_075920.4 | 1 - 2 | 107 | 55 |
GenBank mRNA | AK154610.1 | - | 1 - 2 | 26 | 55 |
AK158750.1 | - | 1 - 2 | 107 | 55 | |
AK159972.1 | - | 1 - 2 | 32 | 55 | |
AK170459.1 | - | 1 - 2 | 26 | 55 |