Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Supv3l1
Entrez Gene ID
338359
Gene Name
suppressor of var1, 3-like 1 (S. cerevisiae)
Gene Aliases
6330443E10Rik
Chromosome Location
Chr.10: 62429329 - 62451294 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181423.2 NP_852088.1 1 - 2 270 96
GenBank mRNA AK031911.1 - 1 - 2 261 96
AK143755.1 - 1 - 2 295 96
BC049796.1 - 1 - 2 270 96

View Full Product Details