The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101482.2 | NP_001094952.2 | 3 - 4 | 71 | |
NM_001177731.1 | NP_001171202.1 | 4 - 5 | 71 | ||
GenBank mRNA | CJ059941.1 | - | 4 - 5 | 71 |