Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mypn
Entrez Gene ID
68802
Gene Name
myopalladin
Gene Aliases
1110056A04Rik, 9830112J03, AI853556, mKIAA4170
Chromosome Location
Chr.10: 63115795 - 63203952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182992.2 NP_892037.2 5 - 6 1475 65
GenBank mRNA AK036458.1 - 5 - 6 1436 65
AK220521.1 - 5 - 6 1475 65

View Full Product Details