The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172920.4 | NP_766508.3 | 13 - 14 | 1327 | 118 |
GenBank mRNA | AK143845.1 | - | 14 - 15 | 1332 | 118 |
BC031382.1 | - | 6 - 7 | 505 | 118 |