Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tprg
Entrez Gene ID
71338
Gene Name
transformation related protein 63 regulated
Gene Aliases
5430420C16Rik, Tprg1
Chromosome Location
Chr.16: 25286817 - 25423588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175165.3 NP_780374.1 1 - 2 211 63
GenBank mRNA AK017323.1 - 1 - 2 207 63
AK036796.1 - 1 - 2 214 63

View Full Product Details