The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291910.1 | NP_001278839.1 | 3 - 4 | 2228 | 73 |
NM_009624.3 | NP_033754.2 | 2 - 3 | 2451 | 73 | |
GenBank mRNA | AK156940.1 | - | 3 - 4 | 2230 | 73 |
BE862471.1 | - | 3 - 4 | 73 | ||
U30602.1 | - | 2 - 3 | 1758 | 73 | |
Z50190.1 | - | 1 - 2 | 1727 | 73 |