Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cluap1
Entrez Gene ID
76779
Gene Name
clusterin associated protein 1
Gene Aliases
2310030D15Rik, 2610111M03Rik
Chromosome Location
Chr.16: 3907659 - 3941147 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029738.2 NP_084014.1 11 - 12 1218 108
GenBank mRNA AK134648.1 - 11 - 12 1185 108
BC024910.1 - 11 - 12 1218 108
BC063769.1 - 5 - 6 452 108

View Full Product Details