Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txndc11
Entrez Gene ID
106200
Gene Name
thioredoxin domain containing 11
Gene Aliases
2810408E11Rik, AI427833, EFP1, Txdc11
Chromosome Location
Chr.16: 11074911 - 11134681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029582.2 NP_083858.1 7 - 8 1119 84
GenBank mRNA AK013044.1 - 7 - 8 1167 84
AK139620.1 - 7 - 8 1201 84
AK155844.1 - 7 - 8 1173 84
AK156154.1 - 7 - 8 1160 84
AK157288.1 - 7 - 8 1093 84
AK157948.1 - 7 - 8 1199 84
AK159484.1 - 7 - 8 1176 84
AK170352.1 - 6 - 7 1047 84
BC020363.1 - 6 - 7 1095 84

View Full Product Details