Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rsl1d1
Entrez Gene ID
66409
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
2410005K20Rik, C76439, pBK1
Chromosome Location
Chr.16: 11193037 - 11203292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025546.2 NP_079822.1 6 - 7 769 89
GenBank mRNA AK012937.1 - 6 - 7 762 89
AK075933.1 - 6 - 7 770 89
AK136025.1 - 6 - 7 736 89
AK145699.1 - 6 - 7 783 89
AK162241.1 - 6 - 7 743 89
AK165572.1 - 6 - 7 782 89
AK166479.1 - 6 - 7 745 89
AK166579.1 - 6 - 7 765 89
AK168469.1 - 6 - 7 765 89
AK168717.1 - 6 - 7 758 89
BC034355.1 - 6 - 7 735 89
BC069934.1 - 6 - 7 743 89
DQ445383.1 - 6 - 7 777 89

View Full Product Details