The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081429.1 | NP_001074898.1 | 8 - 9 | 1914 | 138 |
GenBank mRNA | AK035850.1 | - | 8 - 9 | 1617 | 138 |
AK041803.1 | - | 8 - 9 | 1673 | 138 | |
AK045055.1 | - | 8 - 9 | 1903 | 138 | |
AK133297.1 | - | 8 - 9 | 1880 | 138 | |
AK138231.1 | - | 8 - 9 | 1907 | 138 |