Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajb11
Entrez Gene ID
67838
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 11
Gene Aliases
1810031F23Rik, ABBP-2, AL024055, Dj9, ERdj3, ERj3p
Chromosome Location
Chr.16: 22857845 - 22879634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190804.1 NP_001177733.1 8 - 9 1049 64
NM_001190805.1 NP_001177734.1 8 - 9 1049 64
NM_026400.5 NP_080676.3 8 - 9 1049 64
GenBank mRNA AK008032.1 - 3 - 4 232 64
AK010861.1 - 8 - 9 1017 64
AK050500.1 - 8 - 9 1008 64
AK075785.1 - 8 - 9 1031 64
AK075953.1 - 8 - 9 1011 64
AK148511.1 - 8 - 9 1029 64
AK166730.1 - 8 - 9 1007 64
AY054981.1 - 8 - 9 925 64
BC003999.2 - 8 - 9 980 64
BC018282.1 - 8 - 9 976 64
BC040747.1 - 8 - 9 980 64

View Full Product Details