Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmox2
Entrez Gene ID
15369
Gene Name
heme oxygenase 2
Gene Aliases
HO-2, HO2
Chromosome Location
Chr.16: 4726361 - 4766249 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136066.2 NP_001129538.1 4 - 5 845 89
NM_010443.2 NP_034573.2 4 - 5 861 89
GenBank mRNA AF029874.1 - 4 - 5 858 89
AF054669.1 - 4 - 5 862 89
AF054670.1 - 4 - 5 796 89
AK032175.1 - 4 - 5 803 89
AK134536.1 - 4 - 5 820 89
AK141426.1 - 4 - 5 839 89
AK152936.1 - 4 - 5 784 89
BC002011.1 - 4 - 5 817 89
BP769523.1 - 2 - 3 89

View Full Product Details