The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010676.2 | NP_034806.2 | 1 - 1 | 235 | 156 |
GenBank mRNA | AA717139.1 | - | 1 - 1 | 242 | 156 |
D86422.1 | - | 1 - 1 | 246 | 156 |