Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osbpl11
Entrez Gene ID
106326
Gene Name
oxysterol binding protein-like 11
Gene Aliases
9430097N02Rik, AI132306, AU019795, ORP-11
Chromosome Location
Chr.16: 33169763 - 33243312 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176840.3 NP_789810.2 12 - 13 2946 91
GenBank mRNA AK035188.1 - 2 - 3 329 91
BC035278.1 - 12 - 13 2946 91

View Full Product Details