Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sik2
Entrez Gene ID
235344
Gene Name
salt inducible kinase 2
Gene Aliases
G630080D20Rik, Snf1lk2
Chromosome Location
Chr.9: 50889644 - 51009073 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178710.3 NP_848825.2 13 - 14 2252 61
GenBank mRNA AB067780.1 - 13 - 14 2153 61
AK090386.1 - 13 - 14 2153 61

View Full Product Details