Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfkbiz
Entrez Gene ID
80859
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta
Gene Aliases
AA408868, INAP, Mail
Chromosome Location
Chr.16: 55811377 - 55838641 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159394.1 NP_001152866.1 10 - 11 2045 56
NM_001159395.1 NP_001152867.1 11 - 12 1927 56
NM_030612.3 NP_085115.1 12 - 13 2275 56
GenBank mRNA AB020974.1 - 12 - 13 2487 56
AB026551.2 - 10 - 11 2045 56
AB047549.2 - 11 - 12 1940 56
AK082908.1 - 10 - 11 2044 56
AK136509.1 - 7 - 8 1514 56
AK141318.1 - 10 - 11 2046 56
AK149946.1 - 11 - 12 1923 56
AK150519.1 - 10 - 11 2044 56
AK152016.1 - 10 - 11 2046 56
AK156826.1 - 10 - 11 2045 56
AK159093.1 - 10 - 11 2046 56
AK159171.1 - 10 - 11 2046 56
AK159459.1 - 10 - 11 2046 56
AK159645.1 - 10 - 11 2046 56
AK159917.1 - 10 - 11 2046 56
AK160065.1 - 10 - 11 2044 56
BC058188.1 - 10 - 11 2030 56

View Full Product Details