The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020586.2 | NP_065611.1 | 7 - 8 | 1873 | 76 |
GenBank mRNA | AB041580.1 | - | 7 - 8 | 1668 | 76 |
AK019862.1 | - | 7 - 8 | 1818 | 76 | |
AK135935.1 | - | 7 - 8 | 1874 | 76 | |
AK180080.1 | - | - | 84 | 76 | |
AK208269.1 | - | 1 - 2 | 84 | 76 | |
AK209268.1 | - | 1 - 2 | 84 | 76 | |
AK214130.1 | - | 1 - 2 | 84 | 76 | |
BC029691.1 | - | 7 - 8 | 1091 | 76 | |
BC043693.1 | - | 7 - 8 | 1912 | 76 |