Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Omt2a
Entrez Gene ID
18379
Gene Name
oocyte maturation, alpha
Gene Aliases
OM2a
Chromosome Location
Chr.9: 78311972 - 78314797 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164523.1 NP_001157995.1 1 - 2 101 111
GenBank mRNA AA619817.1 - 1 - 2 90 111
AA693017.1 - 1 - 2 101 111

View Full Product Details