Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmg20a
Entrez Gene ID
66867
Gene Name
high mobility group 20A
Gene Aliases
1200004E06Rik, 5730490E10Rik, Hmgxb1, Ibraf
Chromosome Location
Chr.9: 56418628 - 56496936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025812.2 NP_080088.1 5 - 6 672 86
GenBank mRNA AK004596.1 - 5 - 6 692 86
AK017716.1 - 5 - 6 875 86
AK039222.1 - 5 - 6 694 86
AK129010.1 - 5 - 6 874 86
AK134213.1 - 5 - 6 844 86
BC013804.1 - 5 - 6 672 86
BC068257.1 - 5 - 6 745 86
DQ182735.1 - 5 - 6 672 86

View Full Product Details