Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmg20a
Entrez Gene ID
66867
Gene Name
high mobility group 20A
Gene Aliases
1200004E06Rik, 5730490E10Rik, Hmgxb1, Ibraf
Chromosome Location
Chr.9: 56418628 - 56496936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025812.2 NP_080088.1 8 - 9 987 77
GenBank mRNA AK004596.1 - 8 - 9 1007 77
AK134213.1 - 8 - 9 1159 77
BC013804.1 - 8 - 9 987 77
DQ182735.1 - 8 - 9 987 77

View Full Product Details