Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mknk2
Entrez Gene ID
17347
Gene Name
MAP kinase-interacting serine/threonine kinase 2
Gene Aliases
2010016G11Rik, Gprk7, Mnk2
Chromosome Location
Chr.10: 80665318 - 80676293 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB164081.1 - 6 - 7 495 65
AK030830.1 - 1 - 2 47 65
AK154235.1 - 7 - 8 775 65
BC010256.1 - 5 - 6 405 65
BY732164.1 - 6 - 7 498 65
Y11092.1 - 5 - 6 446 65

View Full Product Details