Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pias4
Entrez Gene ID
59004
Gene Name
protein inhibitor of activated STAT 4
Gene Aliases
PIASY, Pias-gamma, Piasg
Chromosome Location
Chr.10: 81153961 - 81167720 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021501.4 NP_067476.2 8 - 9 968 56
GenBank mRNA AF109174.1 - 8 - 9 1170 56
AK131822.1 - - 971 56
AK131903.1 - 8 - 9 960 56
AK145367.1 - - 1063 56
AK147813.1 - 8 - 9 999 56
AK148066.1 - - 1890 56
AK208285.1 - 2 - 3 184 56
BC020476.1 - 7 - 8 934 56
BC025159.1 - 8 - 9 968 56

View Full Product Details