Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Jsrp1
- Entrez Gene ID
-
71912
- Gene Name
- junctional sarcoplasmic reticulum protein 1
- Gene Aliases
-
2300003C06Rik, 2310032K21Rik, JP-45, JP45
- Chromosome Location
-
Chr.10: 80808494 - 80813593 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK009578.1 |
- |
3 - 4 |
407 |
80 |
View Full Product Details